Learn More
Aviva Systems Biology Corp MRPS2 Antibody - middle region (ARP56813_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56813_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPS2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MRPS2. Alternate names: CGI-91, MRP-S2, S2mtApplicationWB. Species reactivity: Human
Catalog No. 50-286-216