Learn More
ABclonal Technology MRPS31 Rabbit pAb
SDP
Catalog No. 5000098800
Encompass_Preferred
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and aid in protein synthesis within mitochondria. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit, with about 75% protein to rRNA composition, unlike prokaryotic ribosomes, which have a higher rRNA ratio. Unlike prokaryotic ribosomes, mitoribosomes lack a 5S rRNA. The proteins in mammalian mitoribosomes vary greatly across species, complicating sequence homology recognition. The 28S subunit is believed to play a key role in translation initiation. This gene encodes a 28S subunit protein, which has been linked to type 1 diabetes, though its role in the diseases etiology is not fully understood. Pseudogenes corresponding to this gene are found on chromosomes 3 and 13.
Catalog No. 50-000-98800 Supplier ABclonal Technology Supplier No. A1292320UL
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and aid in protein synthesis within mitochondria. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit, with about 75% protein to rRNA composition, unlike prokaryotic ribosomes, which have a higher rRNA ratio. Unlike prokaryotic ribosomes, mitoribosomes lack a 5S rRNA. The proteins in mammalian mitoribosomes vary greatly across species, complicating sequence homology recognition. The 28S subunit is believed to play a key role in translation initiation. This gene encodes a 28S subunit protein, which has been linked to type 1 diabetes, though its role in the diseases etiology is not fully understood. Pseudogenes corresponding to this gene are found on chromosomes 3 and 13.