Learn More
Aviva Systems Biology Corp MRPS6 Antibody - C-terminal region (ARP45882_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45882_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPS6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MRPS6. Alternate names: S6mt, RPMS6, MRP-S6, C21orf101ApplicationWB. Species reactivity: Human
Catalog No. 50-285-224