Learn More
Aviva Systems Biology Corp MRPS9 Antibody - C-terminal region (ARP62694_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62694_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPS9. It was validated on Western Blot. . Alternate names: MRP-S9, RPMS9, S9mtApplicationWB. Species reactivity: Dog, Human, Pig, Rat
Catalog No. 50-287-745