Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRS 1334, Tocris Bioscience™

Catalog No. 138550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-855-0 50 mg
13-851-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-855-0 Supplier R&D Systems Supplier No. 1385/50
This item is not returnable. View return policy

Potent, highly selective hA3 antagonist

Potent and highly selective antagonist for the human adenosine A3 receptor. Ki values are 2.69 nM at hA3, and > 100 μM at rat A1 and rat A2A receptors.

Chemical Identifiers

CAS 192053-05-7
Molecular Formula C31H26N2O6
Molecular Weight (g/mol) 522.557
InChI Key WICWQHSRWZIPNB-AREMUKBSSA-N
Synonym 1, 4-Dihydro-2-methyl-6-phenyl-4-(phenylethynyl)-3, 5-pyridinedicarboxylic acid 3-ethyl-5-[(3-nitrophenyl)methyl] ester
PubChem CID 98072178
IUPAC Name 3-O-ethyl 5-O-[(3-nitrophenyl)methyl] (4R)-2-methyl-6-phenyl-4-(2-phenylethynyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C#CC2=CC=CC=C2)C(=O)OCC3=CC(=CC=C3)[N+](=O)[O-])C4=CC=CC=C4)C

Specifications

Quantity 50 mg
Formula Weight 522.56
Percent Purity >98%
Chemical Name or Material MRS 1334
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.