Learn More
Medchemexpress LLC HY-103186 10mg Medchemexpress, MRS-1706 CAS:264622-53-9 Purity:>98%
SDP
Catalog No. 501968053
Encompass_Preferred
Medchemexpress, HY-103186 10mg MRS-1706 CAS:264622-53-9 MRS-1706 is a potent and selective adenosine A2B receptor inverse agonist. MRS-1706 has Ki values of 1.39, 112, 157, and 230 nM for human A2B, A2A, A1 and A3 receptors respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8053 Supplier Medchemexpress LLC Supplier No. HY10318610MG
Medchemexpress, HY-103186 10mg MRS-1706 CAS:264622-53-9 MRS-1706 is a potent and selective adenosine A2B receptor inverse agonist. MRS-1706 has Ki values of 1.39, 112, 157, and 230 nM for human A2B, A2A, A1 and A3 receptors respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.