Learn More
Medchemexpress LLC MRS 2211 sodium hydrate | 1197030-56-0 | MFCD09038558 | 97.9% | 470.69 g/mol | C14H13ClN4NaO9P | 5 MG
SDP
Catalog No. 5000223490
Encompass_Preferred
MRS 2211 sodium hydrate is the sodium salt hydrate form of a selective competitive P2Y13 receptor antagonist supplied for research use. It is provided as a brown to reddish-brown solid with reported purity of 97.9%, intended for in vitro pharmacology and biochemical studies of purinergic signaling.
  • Selective P2Y13 receptor antagonism (pIC50 ≈ 5.97).
  • High reported purity for reliable experimental results.
  • Solid form suitable for dissolution and formulation in common solvents.
  • Available in small quantities for exploratory and pilot studies.
  • Recommended sealed, cold storage to minimize degradation.
Catalog No. 50-002-23490 Supplier Medchemexpress LLC Supplier No. HY1379555MG
MRS 2211 sodium hydrate is the sodium salt hydrate form of a selective competitive P2Y13 receptor antagonist supplied for research use. It is provided as a brown to reddish-brown solid with reported purity of 97.9%, intended for in vitro pharmacology and biochemical studies of purinergic signaling.
  • Selective P2Y13 receptor antagonism (pIC50 ≈ 5.97).
  • High reported purity for reliable experimental results.
  • Solid form suitable for dissolution and formulation in common solvents.
  • Available in small quantities for exploratory and pilot studies.
  • Recommended sealed, cold storage to minimize degradation.