Learn More
Medchemexpress LLC MRS-3777 hemioxalate | 1186195-57-2 | 96.3% | 399.40 | 50 MG
SDP
Catalog No. 5000344202
Encompass_Preferred
MRS-3777 hemioxalate is a selective adenosine A3 receptor antagonist. It is intended for research use only and is not sold to patients.
  • Selective adenosine A3 receptor antagonist.
  • Available in various quantities for purchase.
  • Documentation available: Data sheet, COA, SDS, handling instructions.
  • Solubility information provided for in vitro and in vivo experiments.
Catalog No. 50-003-44202 Supplier Medchemexpress LLC Supplier No. HY11003750MG
MRS-3777 hemioxalate is a selective adenosine A3 receptor antagonist. It is intended for research use only and is not sold to patients.
  • Selective adenosine A3 receptor antagonist.
  • Available in various quantities for purchase.
  • Documentation available: Data sheet, COA, SDS, handling instructions.
  • Solubility information provided for in vitro and in vivo experiments.