Learn More
Medchemexpress LLC HY-110037 5mg Medchemexpress, MRS-3777 (hemioxalate) CAS:1186195-57-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1100375MG
Encompass_Preferred
Medchemexpress, HY-110037 5mg MRS-3777 (hemioxalate) CAS:1186195-57-2 MRS-3777 hemioxalate is a selective adenosine A3 receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0775