Learn More
MOLTOX Molecular Toxicology Inc Lactobacilli MRS Agar, 500 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948833
Encompass_Preferred
Lactobacilli MRS Agar is used in the isolation, enumeration and cultivation of the Lactobacillus species. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-833 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S071.047
Lactobacilli MRS Agar is used in the isolation, enumeration and cultivation of the Lactobacillus species. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.