Learn More
MOLTOX Molecular Toxicology Inc MRS Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948818
Encompass_Preferred
Used for the isolation of Lactobacillus and Leuconostoc. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-818 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S020.140
Used for the isolation of Lactobacillus and Leuconostoc. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.