Learn More
Teknova MRS Agar Plates, 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. NC0153592
Encompass
MRS media is a selective medium to favor the growth ofLactobacilli.It containssodium acetate, which suppresses the growth of many competing bacteria (although some otherLactobacillales, likeLeuconostocandPediococcus, may grow).
This item is not returnable. View return policy
Catalog No. NC0153592 Supplier Teknova Supplier No. M1350
MRS media is a selective medium to favor the growth ofLactobacilli.It containssodium acetate, which suppresses the growth of many competing bacteria (although some otherLactobacillales, likeLeuconostocandPediococcus, may grow).