Learn More
Medchemexpress LLC HY-103310 10mg Medchemexpress, MRS1845 CAS:544478-19-5 Purity:>98%
SDP
Catalog No. 502080666
Encompass_Preferred
Medchemexpress, HY-103310 10mg MRS1845 CAS:544478-19-5 MRS1845 is a selective store-operated calcium (SOC) channel inhibitor with an IC50 of 1.7 μM. MRS1845 is an ORAI1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0666 Supplier Medchemexpress LLC Supplier No. HY10331010MG
Medchemexpress, HY-103310 10mg MRS1845 CAS:544478-19-5 MRS1845 is a selective store-operated calcium (SOC) channel inhibitor with an IC50 of 1.7 μM. MRS1845 is an ORAI1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.