Learn More
Medchemexpress LLC MRS2279 diammonium | 2387505-47-5 | 98.0% | 503.77 | 50 MG
SDP
Catalog No. 5000346384
Encompass_Preferred
MRS2279 diammonium is a selective, high-affinity P2Y1 receptor antagonist. It competitively inhibits ADP-promoted platelet aggregation. The salt form offers enhanced water solubility and stability, while maintaining comparable biological activity to the free form. This product is intended for research use only.
  • Selective and high-affinity P2Y1 receptor antagonist
  • Competitively inhibits ADP-promoted platelet aggregation
  • Enhanced water solubility
  • Improved stability
Catalog No. 50-003-46384 Supplier Medchemexpress LLC Supplier No. HY108657A1MG
MRS2279 diammonium is a selective, high-affinity P2Y1 receptor antagonist. It competitively inhibits ADP-promoted platelet aggregation. The salt form offers enhanced water solubility and stability, while maintaining comparable biological activity to the free form. This product is intended for research use only.
  • Selective and high-affinity P2Y1 receptor antagonist
  • Competitively inhibits ADP-promoted platelet aggregation
  • Enhanced water solubility
  • Improved stability