Learn More
Medchemexpress LLC MRS2500 tetraammonium | 630103-23-0 | 99.9% | 629.28 | 1 ML
SDP
Catalog No. 5000345248
Encompass_Preferred
MRS2500 tetraammonium is a potent, selective, and stable antagonist of the P2Y1 receptor (Ki=0.78 nM for recombinant human P2Y1 receptor). It inhibits the ADP-induced aggregation of human platelets with an IC50 value of 0.95 nM, demonstrating antithrombotic activity.
  • Potent, selective, and stable antagonist of the P2Y1 receptor.
  • Inhibits ADP-induced aggregation of human platelets with an IC50 of 0.95 nM.
  • Exhibits antithrombotic activity.
  • Decreases acute systemic thromboembolism through selective inhibition of the P2Y1 receptor.
  • Showed strong antithrombotic efficacy in preventing arterial thrombosis in the monkey ECAT model.
Catalog No. 50-003-45248 Supplier Medchemexpress LLC Supplier No. HY1086582MG
MRS2500 tetraammonium is a potent, selective, and stable antagonist of the P2Y1 receptor (Ki=0.78 nM for recombinant human P2Y1 receptor). It inhibits the ADP-induced aggregation of human platelets with an IC50 value of 0.95 nM, demonstrating antithrombotic activity.
  • Potent, selective, and stable antagonist of the P2Y1 receptor.
  • Inhibits ADP-induced aggregation of human platelets with an IC50 of 0.95 nM.
  • Exhibits antithrombotic activity.
  • Decreases acute systemic thromboembolism through selective inhibition of the P2Y1 receptor.
  • Showed strong antithrombotic efficacy in preventing arterial thrombosis in the monkey ECAT model.