Learn More
Medchemexpress LLC MRS2500 tetraammonium | 630103-23-0 | 99.9% | 629.28 | 10 MG
SDP
Catalog No. 5000346161
Encompass_Preferred
MRS2500 tetraammonium is a potent, selective, and stable antagonist of the P2Y1 receptor, with a Ki of 0.78 nM for the recombinant human P2Y1 receptor. It inhibits ADP-induced aggregation of human platelets with an IC50 value of 0.95 nM, exhibiting antithrombotic activity. In vivo studies show it decreases acute systemic thromboembolism through selective inhibition of the P2Y1 receptor and demonstrated strong antithrombotic efficacy in preventing arterial thrombosis.
  • Potent, selective, and stable antagonist of the P2Y1 receptor (K₁=0.78 nM).
  • Inhibits ADP-induced aggregation of human platelets (IC50 of 0.95 nM in human washed platelets).
  • Decreases acute systemic thromboembolism through selective P2Y1 receptor inhibition.
  • Exhibited strong antithrombotic efficacy in preventing arterial thrombosis.
Catalog No. 50-003-46161 Supplier Medchemexpress LLC Supplier No. HY10865810MG
MRS2500 tetraammonium is a potent, selective, and stable antagonist of the P2Y1 receptor, with a Ki of 0.78 nM for the recombinant human P2Y1 receptor. It inhibits ADP-induced aggregation of human platelets with an IC50 value of 0.95 nM, exhibiting antithrombotic activity. In vivo studies show it decreases acute systemic thromboembolism through selective inhibition of the P2Y1 receptor and demonstrated strong antithrombotic efficacy in preventing arterial thrombosis.
  • Potent, selective, and stable antagonist of the P2Y1 receptor (K₁=0.78 nM).
  • Inhibits ADP-induced aggregation of human platelets (IC50 of 0.95 nM in human washed platelets).
  • Decreases acute systemic thromboembolism through selective P2Y1 receptor inhibition.
  • Exhibited strong antithrombotic efficacy in preventing arterial thrombosis.