Learn More
Medchemexpress LLC MRS2500 tetraammonium | 630103-23-0 | 99.9% | 629.28 g·mol⁻¹ | C13H30IN9O8P2 | 5 MG
SDP
Catalog No. 5000263509
Encompass_Preferred
MRS2500 tetraammonium is a research-grade tetraammonium salt and a potent, selective antagonist of the P2Y1 receptor. It inhibits ADP-induced human platelet aggregation at sub-nanomolar concentrations and is supplied in small, high-purity quantities for pharmacology and platelet-aggregation research. The compound is water soluble and suitable for in vitro assays.
Catalog No. 50-002-63509 Supplier Medchemexpress LLC Supplier No. HY1086585MG
MRS2500 tetraammonium is a research-grade tetraammonium salt and a potent, selective antagonist of the P2Y1 receptor. It inhibits ADP-induced human platelet aggregation at sub-nanomolar concentrations and is supplied in small, high-purity quantities for pharmacology and platelet-aggregation research. The compound is water soluble and suitable for in vitro assays.