Learn More
Aviva Systems Biology Corp MRS2L Antibody - middle region (ARP49536_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49536_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRS2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MRS2L. Alternate names: HPT, MGC78523, MRS2, MRS2LApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60107