Learn More
Cayman Chemical MRSA Antibiotic 1, Purity: ≥95%, CAS: 667410-81-3, 10 mg
SDP
Catalog No. 502943984
Encompass_Preferred
An antibiotic; active against S. aureus, methicillin-resistant S. aureus (MRSA), and B. subtilis (MICs = 2, 4, and 2 µg/ml, respectively) without inducing cytotoxicity in HepG2, HSkMC, or IMR-90 cells at 10 µM; active against 40 antibiotic-resistant clinical isolates (median MICs = 2-16 µg/ml); decreases MRSA viral titers in the skin in a neutropenic mouse wound infection model when applied topically at 1% (w/w); reduces MRSA viral titers in the thigh in a neutropenic mouse thigh infection model at 80 mg/kg i.p.
Catalog No. 50-294-3984 Supplier Cayman Chemical Supplier No. 4043210 MG
An antibiotic; active against S. aureus, methicillin-resistant S. aureus (MRSA), and B. subtilis (MICs = 2, 4, and 2 µg/ml, respectively) without inducing cytotoxicity in HepG2, HSkMC, or IMR-90 cells at 10 µM; active against 40 antibiotic-resistant clinical isolates (median MICs = 2-16 µg/ml); decreases MRSA viral titers in the skin in a neutropenic mouse wound infection model when applied topically at 1% (w/w); reduces MRSA viral titers in the thigh in a neutropenic mouse thigh infection model at 80 mg/kg i.p.