Learn More
Cayman Chemical MRSA ntIbIotIc 2 10mg
SDP
Catalog No. 502945386
Encompass_Preferred
An antibiotic; active against the Gram-positive bacteria B. subtilis, S. aureus, and MRSA (MICs = 2, 2, and 4 UG/ml, respectively) but not the Gram-negative bacteria E. coli, A. baumannii, and P. aeruginosa (MICs = >128 UG/ml for all); disrupts the pH gradient of S. aureus and B. subtilis in a proton motive force assay at 32 UG/ml
Catalog No. 50-294-5386 Supplier Cayman Chemical Supplier No. 4043310 MG
An antibiotic; active against the Gram-positive bacteria B. subtilis, S. aureus, and MRSA (MICs = 2, 2, and 4 UG/ml, respectively) but not the Gram-negative bacteria E. coli, A. baumannii, and P. aeruginosa (MICs = >128 UG/ml for all); disrupts the pH gradient of S. aureus and B. subtilis in a proton motive force assay at 32 UG/ml