Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ MRSA Screen Plate, Mueller Hinton Agar with Sodium Chloride and Oxacillin
SDP

Catalog No. 23001772 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-772 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-772 Supplier Hardy Diagnostics™ Supplier No. G47
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the detection of Methicillin Resistant Staphylococcus aureus

  • Store at 2 to 8°C away from direct light
  • Use for antibiotic susceptibility testing

Specifications

Appearance Clear, Slightly Opalescent
Color White to Beige
Content And Storage 2°C to 8°C
Country of Origin United States
Additive 4% Sodium Chloride, Oxacillin
Detects Methicillin-resistant Staphylococcus aureus
Form Agar
Format Plate
Manufacturing Origin US with Globally Sourced Materials
Media Type Mueller Hinton Agar with 4% Sodium Chloride and 6mcg/ml of Oxacillin
Product Type Agar Plate
Target Organism Class Gram-positive Cocci
Test Type Antibiotic Susceptibility Sensitivity
Fill Volume 18 mL
Formulation Beef Infusion - 300.0gm, Sodium Chloride - 40.0gm, Casamino Acids - 17.5gm, Starch - 1.5gm, Oxacillin - 6.0mg, Agar - 17.0gm
For Use With (Application) Screening Staphylococcus aureus isolates for resistance to methicillin, nafcillin and oxacillin
Incubation Conditions 35°C
Incubation Time 24 hr.
No. of Places 1
No. per Pack 10
Packaging Type Plates, Bag
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 100 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Staphylococcus species
Protein Source Beef Infusion, Casamino Acids
Source Beef, Casein
Manufacturing Quality ISO 13485, CLSI Document M22-A3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.