Learn More
Cayman Chemical MRSCoV NucleocapsId ProteIn
SDP
Catalog No. 502945711
Encompass_Preferred
Source: Recombinant MERS-CoV C-Terminal His-tagged nucleocapsid protein expressed in insect cells • Amino acids: 1-413 (full length) • MW: 46.51 kDa
Catalog No. 50-294-5711 Supplier Cayman Chemical Supplier No. 40878100 UG
Source: Recombinant MERS-CoV C-Terminal His-tagged nucleocapsid protein expressed in insect cells • Amino acids: 1-413 (full length) • MW: 46.51 kDa