Learn More
Cayman Chemical MRSCoV SpIke GlycoproteIn R
SDP
Catalog No. 502915865
Encompass_Preferred
Immunogen: Recombinant MERS-CoV spike glycoprotein • Host: Rabbit • Species Reactivity: (+) MERS-CoV • Cross Reactivity: (+) MERS-CoV spike glycoprotein, MERS-CoV spike glycoprotein S1, MERS-CoV spike glycoprotein RBD; (-) MERS-CoV spike glycoprotein S2 • Application: MN
Catalog No. 50-291-5865 Supplier Cayman Chemical Supplier No. 38105200 UG
Immunogen: Recombinant MERS-CoV spike glycoprotein • Host: Rabbit • Species Reactivity: (+) MERS-CoV • Cross Reactivity: (+) MERS-CoV spike glycoprotein, MERS-CoV spike glycoprotein S1, MERS-CoV spike glycoprotein RBD; (-) MERS-CoV spike glycoprotein S2 • Application: MN