Learn More
Cayman Chemical MRSCoV SpIke GlycoproteIn R
SDP
Supplier:  Cayman Chemical 40876100 UG
Encompass_Preferred
Source: Recombinant MERS-CoV C-terminal His-tagged spike glycoprotein RBD expressed in insect cells • AA: 367-606 • Tag: C-terminal His • MW: 27.7 kDa
Catalog No. 50-294-4689