Learn More
Uline Marsh Paint Markers | Black | 1 EA
Catalog No. NC2270767
Encompass
General purpose alcohol-based paint resists UV rays, Permanently marks metal wood cardboard and plastic, Weatherproof and waterproof, Contains 12 fluid ounce, Dries in 3 to 5 seconds depending on surface, Tip Valve action controls ink flow, Flammable paint, Ink Toxic, Fades with time in sunlight but pigmented so will last, Removes with thinner but may leave residue
Catalog No. NC2270767 Supplier Uline Supplier No. H257BL
General purpose alcohol-based paint resists UV rays, Permanently marks metal wood cardboard and plastic, Weatherproof and waterproof, Contains 12 fluid ounce, Dries in 3 to 5 seconds depending on surface, Tip Valve action controls ink flow, Flammable paint, Ink Toxic, Fades with time in sunlight but pigmented so will last, Removes with thinner but may leave residue