Learn More
Selleck Chemical LLC MRT-2359-E1427-5MG
SDP
Catalog No. 503137105
Encompass_Preferred
MRT-2359 is a potent degrader of GSPT1 It exhibits anti-tumor activity in non-small cell lung cancer (NSCLC) and small cell lung cancer (SCLC) cells
Catalog No. 50-313-7105 Supplier Selleck Chemical LLC Supplier No. E14275MG
MRT-2359 is a potent degrader of GSPT1 It exhibits anti-tumor activity in non-small cell lung cancer (NSCLC) and small cell lung cancer (SCLC) cells