Learn More
TARGETMOL CHEMICALS INC MRT 67307 DIHYDROCHLORID 5MG
SDP
Catalog No. 502580154
Encompass_Preferred
Also available in 1 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MRT 67307 dihydrochloride is a dual inhibitor of the IKK(epsilon) and TBK-1 with IC50s of 160 and 19 nM respectively. MRT67307 dihydrochloride also inhibits ULK1 and ULK2 with IC50s of 45 and 38 nM respectively. MRT67307 dihydrochloride also blocks autophagy in cells. purity: 98%
Catalog No. 50-258-0154 Supplier TARGETMOL CHEMICALS INC Supplier No. T369945MG
Also available in 1 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MRT 67307 dihydrochloride is a dual inhibitor of the IKK(epsilon) and TBK-1 with IC50s of 160 and 19 nM respectively. MRT67307 dihydrochloride also inhibits ULK1 and ULK2 with IC50s of 45 and 38 nM respectively. MRT67307 dihydrochloride also blocks autophagy in cells. purity: 98%