Learn More
Medchemexpress LLC MRT00033659 | 1401731-54-1 | 99.8% | 266.30 g·mol⁻1 | C15H14N4O | 5 MG
SDP
Catalog No. 5000226823
Encompass_Preferred
Small-molecule kinase inhibitor active against CK1 and CHK1, provided as a high-purity solid for research use.
  • High purity: 99.77% suitable for biochemical and cellular assays.
  • Potent inhibition of CK1 and CHK1 kinases, useful for pathway modulation studies.
  • Provided as a 5 MG solid for small-scale research experiments.
  • Molecular weight 266.30 g·mol⁻1 and formula C15H14N4O.
  • Available in multiple small pack sizes for dose-ranging and pilot studies.
Catalog No. 50-002-26823 Supplier Medchemexpress LLC Supplier No. HY1178575MG
Small-molecule kinase inhibitor active against CK1 and CHK1, provided as a high-purity solid for research use.
  • High purity: 99.77% suitable for biochemical and cellular assays.
  • Potent inhibition of CK1 and CHK1 kinases, useful for pathway modulation studies.
  • Provided as a 5 MG solid for small-scale research experiments.
  • Molecular weight 266.30 g·mol⁻1 and formula C15H14N4O.
  • Available in multiple small pack sizes for dose-ranging and pilot studies.