Learn More
Medchemexpress LLC HY-13018 10mg Medchemexpress, MRT67307 CAS:1190378-57-4 Purity:>98%
SDP
Catalog No. 502028944
Encompass_Preferred
Medchemexpress, HY-13018 10mg MRT67307 CAS:1190378-57-4 MRT67307 is a dual inhibitor of the IKKε and TBK-1 with IC50s of 160 and 19 nM, respectively. MRT67307 also inhibits ULK1 and ULK2 with IC50s of 45 and 38 nM, respectively. MRT67307 also blocks autophagy in cells[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8944 Supplier Medchemexpress LLC Supplier No. HY1301810MG
Medchemexpress, HY-13018 10mg MRT67307 CAS:1190378-57-4 MRT67307 is a dual inhibitor of the IKKε and TBK-1 with IC50s of 160 and 19 nM, respectively. MRT67307 also inhibits ULK1 and ULK2 with IC50s of 45 and 38 nM, respectively. MRT67307 also blocks autophagy in cells[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.