Learn More
Medchemexpress LLC Mrt67307 | 1190378-57-4 | 99.6% | 100 MG
SDP
Catalog No. 5000339043
Encompass_Preferred
MRT67307 is a dual inhibitor of IKKε and TBK-1, with IC50s of 160 nM and 19 nM respectively. It also inhibits ULK1 and ULK2, and blocks autophagy in cells.
  • Inhibits IKKε with an IC50 of 160 nM
  • Inhibits TBK-1 with an IC50 of 19 nM
  • Inhibits ULK1 with an IC50 of 45 nM
  • Inhibits ULK2 with an IC50 of 38 nM
  • Blocks autophagy in cells
Catalog No. 50-003-39043 Supplier Medchemexpress LLC Supplier No. HY13018100MG
MRT67307 is a dual inhibitor of IKKε and TBK-1, with IC50s of 160 nM and 19 nM respectively. It also inhibits ULK1 and ULK2, and blocks autophagy in cells.
  • Inhibits IKKε with an IC50 of 160 nM
  • Inhibits TBK-1 with an IC50 of 19 nM
  • Inhibits ULK1 with an IC50 of 45 nM
  • Inhibits ULK2 with an IC50 of 38 nM
  • Blocks autophagy in cells