Learn More
Apexbio Technology LLC MRT67307 HCl 2095432-39-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063196
Encompass_Preferred
MRT67307 HCl (CAS 2095432-39-4) is a small-molecule inhibitor targeting several kinases including TBK1 IKK MARK1-4 NUAK1 ULK1/2 and salt-inducible kinases (SIK1-3) It inhibits TBK1 and IKK to block phosphorylation of IRF3 reducing interferon- synthesis without affecting NF- B activation Additionally MRT67307-mediated SIK inhibition promotes regulatory macrophage differentiation leading to elevated anti-inflammatory cytokines (e g IL-10 IL-1ra) and decreased pro-inflammatory cytokines (IL-6 IL-12 TNF) It is utilized to investigate innate immunity macrophage polarization autophagy and inflammation pathways
Catalog No. 50-000-63196 Supplier Apexbio Technology LLC Supplier No. A36355.1
MRT67307 HCl (CAS 2095432-39-4) is a small-molecule inhibitor targeting several kinases including TBK1 IKK MARK1-4 NUAK1 ULK1/2 and salt-inducible kinases (SIK1-3) It inhibits TBK1 and IKK to block phosphorylation of IRF3 reducing interferon- synthesis without affecting NF- B activation Additionally MRT67307-mediated SIK inhibition promotes regulatory macrophage differentiation leading to elevated anti-inflammatory cytokines (e g IL-10 IL-1ra) and decreased pro-inflammatory cytokines (IL-6 IL-12 TNF) It is utilized to investigate innate immunity macrophage polarization autophagy and inflammation pathways