Learn More
Selleck Chemical LLC MRT67307 HCl
SDP
Catalog No. 5000000179
Encompass_Preferred
MRT67307 is a potent and dual IKK and TBK1 inhibitor with IC50 of 160 and 19 nM respectively MRT67307 potently inhibits ULK1 and ULK2 and blocks autophagy
Catalog No. 50-000-00179 Supplier Selleck Chemical LLC Supplier No. S79481G
Add to Cart
Add to Cart
MRT67307 is a potent and dual IKK and TBK1 inhibitor with IC50 of 160 and 19 nM respectively MRT67307 potently inhibits ULK1 and ULK2 and blocks autophagy