Learn More
Selleck Chemical LLC MRT68921 HCl 5mg 1190379-70-4(freebase)
SDP
Supplier:  Selleck Chemical LLC S79495MG
Encompass_Preferred
MRT68921 is a potent and dual autophagy kinase ULK1/2 inhibitor with IC50 of 2.9 nM and 1.1 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1200846