Learn More
Medchemexpress LLC HY-100006A 100mg Medchemexpress, MRT68921 (dihydrochloride) CAS:2080306-21-2 Purity:>98%
SDP
Catalog No. 502028598
Encompass_Preferred
Medchemexpress, HY-100006A 100mg MRT68921 (dihydrochloride) CAS:2080306-21-2 MRT68921 dihydrochloride is a potent inhibitor of ULK1 and ULK2, with IC50 values of 2.9 nM and 1.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8598 Supplier Medchemexpress LLC Supplier No. HY100006A100MG
Medchemexpress, HY-100006A 100mg MRT68921 (dihydrochloride) CAS:2080306-21-2 MRT68921 dihydrochloride is a potent inhibitor of ULK1 and ULK2, with IC50 values of 2.9 nM and 1.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.