Learn More
Medchemexpress LLC MRT68921 dihydrochloride | 2080306-21-2 | 99.7% | 507.50 | C25H36Cl2N6O | 10 MG
SDP
Catalog No. 5000215011
Encompass_Preferred
MRT68921 dihydrochloride is a research reagent and potent small-molecule inhibitor of NUAK1 and ULK1 used in cellular studies of autophagy, oxidative stress, and apoptosis. It demonstrates low-nanomolar inhibitory activity against ULK kinases and is used to probe autophagy-related pathways and anti-proliferative responses in cancer research.
  • Potent dual NUAK1/ULK1 inhibitory activity with low-nanomolar IC50 values.
  • Demonstrated ULK2 inhibition at 1.1 nM and ULK1 inhibition at 2.9 nM.
  • Used for in vitro studies of autophagy modulation, ROS production, and apoptosis.
  • High reported purity suitable for analytical and biological assays.
  • Provided as a solid with molecular weight 507.50 and formula C25H36Cl2N6O.
Catalog No. 50-002-15011 Supplier Medchemexpress LLC Supplier No. HY100006A10MG
MRT68921 dihydrochloride is a research reagent and potent small-molecule inhibitor of NUAK1 and ULK1 used in cellular studies of autophagy, oxidative stress, and apoptosis. It demonstrates low-nanomolar inhibitory activity against ULK kinases and is used to probe autophagy-related pathways and anti-proliferative responses in cancer research.
  • Potent dual NUAK1/ULK1 inhibitory activity with low-nanomolar IC50 values.
  • Demonstrated ULK2 inhibition at 1.1 nM and ULK1 inhibition at 2.9 nM.
  • Used for in vitro studies of autophagy modulation, ROS production, and apoptosis.
  • High reported purity suitable for analytical and biological assays.
  • Provided as a solid with molecular weight 507.50 and formula C25H36Cl2N6O.