Learn More
Medchemexpress LLC MRT68921 dihydrochloride | 2080306-21-2 | 99.7% | 507.50 g·mol^-1 | C25H36Cl2N6O | 1 MG
SDP
Catalog No. 5000215527
Encompass_Preferred
MRT68921 dihydrochloride is a potent dual NUAK1/ULK1 kinase inhibitor used in cancer and autophagy research. It inhibits ULK1 and ULK2 at low nanomolar concentrations, disrupts autophagy, and promotes apoptosis; the compound is supplied as a solid with documented solubility and storage recommendations for research use.
  • Inhibits ULK1 and ULK2 at low nanomolar potency.
  • Blocks cellular autophagy and promotes apoptosis.
  • High reported purity suitable for biochemical assays.
  • Soluble in water and DMSO with specified preparation conditions.
  • Stable when stored sealed at recommended temperatures.
  • Supplied in small solid package sizes for research use.
Catalog No. 50-002-15527 Supplier Medchemexpress LLC Supplier No. HY100006A1MG
MRT68921 dihydrochloride is a potent dual NUAK1/ULK1 kinase inhibitor used in cancer and autophagy research. It inhibits ULK1 and ULK2 at low nanomolar concentrations, disrupts autophagy, and promotes apoptosis; the compound is supplied as a solid with documented solubility and storage recommendations for research use.
  • Inhibits ULK1 and ULK2 at low nanomolar potency.
  • Blocks cellular autophagy and promotes apoptosis.
  • High reported purity suitable for biochemical assays.
  • Soluble in water and DMSO with specified preparation conditions.
  • Stable when stored sealed at recommended temperatures.
  • Supplied in small solid package sizes for research use.