Learn More
TARGETMOL CHEMICALS INC MRT68921 dihydrochloride 25MG
SDP
Catalog No. 502878044
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. MRT68921 dihydrochloride is a potent ULK1 and ULK2 inhibitor (IC50 of 2.9 nM and 1.1 nM, respectively). Purity 98.98%
Catalog No. 50-287-8044 Supplier TARGETMOL CHEMICALS INC Supplier No. T914225MG
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. MRT68921 dihydrochloride is a potent ULK1 and ULK2 inhibitor (IC50 of 2.9 nM and 1.1 nM, respectively). Purity 98.98%