Learn More
Selleck Chemical LLC MRT68921 HCl
SDP
Supplier:  Selleck Chemical LLC S7949100MG
Encompass_Preferred
MRT68921 is a potent and dual autophagy kinase ULK1/2 inhibitor with IC50 of 2 9 nM and 1 1 nM respectively
Catalog No. 50-000-00170