Learn More
Cayman Chemical MRTK EXrAClular DomaInh 10ug
SDP
Catalog No. 502946561
Encompass_Preferred
Source: Active recombinant human N-terminal GST-tagged MERTK expressed in insect cells • Amino acids: 578-872 • MW: 60 kDa
Catalog No. 50-294-6561 Supplier Cayman Chemical Supplier No. 3374310 UG
Source: Active recombinant human N-terminal GST-tagged MERTK expressed in insect cells • Amino acids: 578-872 • MW: 60 kDa