Learn More
Aviva Systems Biology Corp MRTO4 Antibody - middle region (ARP46239_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46239_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRTO4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MRTO4. Alternate names: C1orf33, MRT4, dJ657E11.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-245