Learn More
Medchemexpress LLC HY-114436 10mg Medchemexpress, MRTX-1257 CAS:2206736-04-9 Purity:>98%
SDP
Catalog No. 501969358
Encompass_Preferred
Medchemexpress, HY-114436 10mg MRTX-1257 CAS:2206736-04-9 MRTX-1257 is a selective, irreversible, covalent and oral active KRAS G12C inhibitor, with an IC50 of 900 pM for KRAS dependent ERK phosphorylation in H358 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9358 Supplier Medchemexpress LLC Supplier No. HY11443610MG
Medchemexpress, HY-114436 10mg MRTX-1257 CAS:2206736-04-9 MRTX-1257 is a selective, irreversible, covalent and oral active KRAS G12C inhibitor, with an IC50 of 900 pM for KRAS dependent ERK phosphorylation in H358 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.