Learn More
Medchemexpress LLC HY-114436 5mg Medchemexpress, MRTX-1257 CAS:2206736-04-9 Purity:>98%
SDP
Catalog No. 501932493
Encompass_Preferred
Medchemexpress, HY-114436 5mg MRTX-1257 CAS:2206736-04-9 MRTX-1257 is a selective, irreversible, covalent and oral active KRAS G12C inhibitor, with an IC 50 of 900 pM for KRAS dependent ERK phosphorylation in H358 cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2493 Supplier Medchemexpress LLC Supplier No. HY1144365MG
Medchemexpress, HY-114436 5mg MRTX-1257 CAS:2206736-04-9 MRTX-1257 is a selective, irreversible, covalent and oral active KRAS G12C inhibitor, with an IC 50 of 900 pM for KRAS dependent ERK phosphorylation in H358 cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.