Learn More
TARGETMOL CHEMICALS INC MRTX-1719 1MG
SDP
Catalog No. 502877165
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MRTX-1719 is a potent and selective inhibitor of the PRMT5/MTA complex with an IC50 value of <10 nM against PRMT5/MTAMTAPDELSDMA cell lines. Purity 99.18%
Catalog No. 50-287-7165 Supplier TARGETMOL CHEMICALS INC Supplier No. T402541MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MRTX-1719 is a potent and selective inhibitor of the PRMT5/MTA complex with an IC50 value of <10 nM against PRMT5/MTAMTAPDELSDMA cell lines. Purity 99.18%