Learn More
Medchemexpress LLC HY-139611 5mg , MRTX-1719 CAS:2630904-45-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1396115MG
Encompass_Preferred
HY-139611 5mg MRTX-1719 CAS: 2630904-45-7 MRTX-1719 is a potent first-in-class selective inhibitor of the PRMT5/MTA complex, with an IC50 of less than 10 nM in PRMT5/MTA MTAPDEL SDMA cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0123