Learn More
Medchemexpress LLC MRTX-EX185 formic | 2988148-70-3 | 97.2% | 610.68 g/mol | C34H35FN6O4 | 5 MG
SDP
Catalog No. 5000221064
Encompass_Preferred
MRTX-EX185 formic is a small-molecule KRAS(G12D) inhibitor for research use. It binds both GDP-loaded and active GNP states of KRAS and shows biochemical activity (IC50 ≈ 90 nM). The product is supplied as the formic acid salt and is intended for in vitro and in vivo research studies.
Catalog No. 50-002-21064 Supplier Medchemexpress LLC Supplier No. HY145962A5MG
MRTX-EX185 formic is a small-molecule KRAS(G12D) inhibitor for research use. It binds both GDP-loaded and active GNP states of KRAS and shows biochemical activity (IC50 ≈ 90 nM). The product is supplied as the formic acid salt and is intended for in vitro and in vivo research studies.