Learn More
Medchemexpress LLC MRTX-EX185 (formic) | 2988148-70-3 | 97.2% | C34H35FN6O4 | 10MG
SDP
Catalog No. 5000258414
Encompass_Preferred
MRTX-EX185 (formic) is a small-molecule inhibitor of KRAS with potent activity against the oncogenic G12D mutant (IC50 = 90 nM). It binds both GDP-loaded and GNP (GTP-mimetic) states of KRAS and is intended for biochemical, cellular, and tumor-model research. The compound is supplied as a purified solid (yellow to brown), with reported purity around 97.2%, molecular formula C34H35FN6O4, and molecular weight 610.68 g·mol⁻¹.
  • Potent KRAS G12D inhibition (IC50 = 90 nM).
  • Binds both GDP-loaded and GTP-mimetic states of KRAS.
  • Applicable to in vitro and in vivo research studies.
  • High reported purity, suitable for research use.
  • Readily soluble in DMSO (125 mg/mL) for assay preparation.
  • Stable as powder when stored at -20°C for long-term storage.
Catalog No. 50-002-58414 Supplier Medchemexpress LLC Supplier No. HY145962A10MG
MRTX-EX185 (formic) is a small-molecule inhibitor of KRAS with potent activity against the oncogenic G12D mutant (IC50 = 90 nM). It binds both GDP-loaded and GNP (GTP-mimetic) states of KRAS and is intended for biochemical, cellular, and tumor-model research. The compound is supplied as a purified solid (yellow to brown), with reported purity around 97.2%, molecular formula C34H35FN6O4, and molecular weight 610.68 g·mol⁻¹.
  • Potent KRAS G12D inhibition (IC50 = 90 nM).
  • Binds both GDP-loaded and GTP-mimetic states of KRAS.
  • Applicable to in vitro and in vivo research studies.
  • High reported purity, suitable for research use.
  • Readily soluble in DMSO (125 mg/mL) for assay preparation.
  • Stable as powder when stored at -20°C for long-term storage.