Learn More
TARGETMOL CHEMICALS INC MRTX0902 5MG
SDP
Catalog No. 502578964
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. MRTX0902 is an effective and high selective inhibitor of SOS1 with an IC50 of 46 nM and a Ki of 2 nM. purity: 98%
Catalog No. 50-257-8964 Supplier TARGETMOL CHEMICALS INC Supplier No. T97555MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. MRTX0902 is an effective and high selective inhibitor of SOS1 with an IC50 of 46 nM and a Ki of 2 nM. purity: 98%