Learn More
Selleck Chemical LLC MRTX1133-E1051-1MG
SDP
Supplier:  Selleck Chemical LLC E10511MG
Encompass_Preferred
MRTX1133 is a highly selective inhibitor of mutant KRAS G12D and can reversibly binds to the activated and inactivated KRAS G12D mutants and inhibit their activity The specificity of MRTX1133 to KRAS G12D is more than 1000 times that of wild-type KRAS
Catalog No. 50-238-9788