Learn More
Medchemexpress LLC HY-130149 10mg Medchemexpress, MRTX849 CAS:2326521-71-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13014910MG
Encompass_Preferred
Medchemexpress, HY-130149 10mg MRTX849 CAS:2326521-71-3 MRTX849 is a potent, orally-available, and mutation-selective covalent inhibitor of KRAS G12C with potential antineoplastic activity. MRTX849 covalently binds to KRAS G12C at the cysteine at residue 12, locks the protein in its inactive GDP-bound conformation, and inhibits KRAS-dependent signal transduction[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8941