Learn More
Medchemexpress LLC MRTX9768 hydrochloride | 2629314-68-5 | 99.9% | 460.89 | 100 MG
SDP
Catalog No. 5000350055
Encompass_Preferred
MRTX9768 hydrochloride is a potent, selective, orally active, first-in-class PRMT5-MTA complex inhibitor. It inhibits SDMA and cell proliferation in HCT116 MTAP-del cells, showing marked selectivity over HCT116 MTAP-WT cells. This compound leads to sustained SDMA inhibition and demonstrates dose-dependent inhibition in MTAP-del tumors in vivo. It selectively targets MTAP/CDKN2A-deleted tumors, such as glioblastoma, and exhibits a favorable ADME profile.
  • Potent and selective PRMT5-MTA complex inhibitor
  • Orally active
  • Targets MTAP/CDKN2A-deleted tumors
  • Exhibits favorable ADME profile
  • Leads to sustained SDMA inhibition
  • Inhibits cell proliferation in HCT116 MTAP-del cells
Catalog No. 50-003-50055 Supplier Medchemexpress LLC Supplier No. HY138684A100MG
MRTX9768 hydrochloride is a potent, selective, orally active, first-in-class PRMT5-MTA complex inhibitor. It inhibits SDMA and cell proliferation in HCT116 MTAP-del cells, showing marked selectivity over HCT116 MTAP-WT cells. This compound leads to sustained SDMA inhibition and demonstrates dose-dependent inhibition in MTAP-del tumors in vivo. It selectively targets MTAP/CDKN2A-deleted tumors, such as glioblastoma, and exhibits a favorable ADME profile.
  • Potent and selective PRMT5-MTA complex inhibitor
  • Orally active
  • Targets MTAP/CDKN2A-deleted tumors
  • Exhibits favorable ADME profile
  • Leads to sustained SDMA inhibition
  • Inhibits cell proliferation in HCT116 MTAP-del cells